Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naglt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176080.2 | NP_788269.1 | 4 | 619 | ||
GenBank mRNA | AB089802.1 | - | 4 | 619 | ||
BC098651.1 | - | 4 | 606 | 7130354 |
Gene Symbol: RGD1304770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134547.1 | NP_001128019.1 | 4 | 541 |
Gene Symbol: RGD1559971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256535.2 | XP_006256597.1 | 4 | 680 |