Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181090.2 | NP_851604.1 | 5, 6 | 787 | ||
XM_006242282.1 | XP_006242344.1 | 5, 6 | 827 | |||
XM_006242283.1 | XP_006242345.1 | 4, 5 | 711 | |||
GenBank mRNA | AF173682.2 | - | 5, 6 | 588 | ||
AF249673.1 | - | 5, 6 | 787 | |||
AF273024.1 | - | 5, 6 | 622 |