Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181091.2 | NP_851605.1 | 3, 4 | 213 | ||
GenBank mRNA | AB007364.1 | - | 3, 4 | 213 |