Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037632.1 | NP_001032721.1 | 2 | 353 | ||
NM_181374.2 | NP_852039.2 | 2 | 233 | |||
XM_006242390.2 | XP_006242452.1 | 2 | 589 | |||
XM_008765706.1 | XP_008763928.1 | 3 | 159 | |||
GenBank mRNA | AY177399.1 | - | 2 | 135 |