Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181381.2 | NP_852046.1 | 3, 4 | 339 | ||
XM_006236572.2 | XP_006236634.1 | 3, 4 | 376 | |||
XM_006236573.2 | XP_006236635.1 | 3, 4 | 510 | |||
XM_006236574.2 | XP_006236636.1 | 3, 4 | 430 | |||
XM_006236575.2 | XP_006236637.1 | 3, 4 | 1030 | |||
XM_006236576.1 | XP_006236638.1 | 3, 4 | 441 | |||
XM_006236577.2 | XP_006236639.1 | 2, 3 | 341 | |||
XM_008762961.1 | XP_008761183.1 | 3, 4 | 400 | |||
GenBank mRNA | AB094089.1 | - | 2, 3 | 258 | ||
AB105817.1 | - | 3, 4 | 313 | |||
AY089996.1 | - | 3, 4 | 339 | |||
AY089997.1 | - | 3, 4 | 339 | |||
AY089998.1 | - | 3, 4 | 339 |