Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178333.1 | NP_848016.1 | 1, 2 | 121 | ||
XM_008773069.1 | XP_008771291.1 | 3, 4 | 1451 | |||
XM_008773070.1 | XP_008771292.1 | 3, 4 | 1451 | |||
GenBank mRNA | AB098162.1 | - | 1, 2 | 121 |