Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181473.2 | NP_852138.2 | 3 | 1132 | ||
GenBank mRNA | AB101615.1 | - | 2 | 984 | ||
BC092204.1 | - | 3 | 1132 | 7312023 | ||
FQ219171.1 | - | 4 | 1215 |