Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012659.1 | NP_036791.1 | 1 | 157 | ||
GenBank mRNA | M25890.1 | - | 1 | 157 | ||
V01271.1 | - | 1 | 157 |