Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012674.2 | NP_036806.1 | 2 | 135 | ||
GenBank mRNA | M22162.1 | - | 2 | 75 | ||
M27882.1 | - | 2 | 117 | |||
M35299.1 | - | 84 | ||||
X59696.1 | - | 2 | 135 |