Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013226.2 | NP_037358.1 | 2 | 135 | ||
GenBank mRNA | BC061562.1 | - | 2 | 135 | 6921029 | |
BC082797.1 | - | 2 | 120 | 7301263 | ||
FQ209939.1 | - | 2 | 131 | |||
FQ210972.1 | - | 2 | 128 | |||
FQ211501.1 | - | 2 | 128 | |||
FQ211742.1 | - | 2 | 132 | |||
FQ212686.1 | - | 2 | 128 | |||
FQ216977.1 | - | 2 | 131 | |||
FQ216988.1 | - | 2 | 131 | |||
FQ217307.1 | - | 2 | 131 | |||
FQ217782.1 | - | 2 | 131 | |||
FQ217854.1 | - | 2 | 131 | |||
FQ219795.1 | - | 2 | 128 | |||
FQ220681.1 | - | 2 | 129 | |||
FQ221405.1 | - | 1 | 97 | |||
FQ221557.1 | - | 2 | 128 | |||
FQ221558.1 | - | 2 | 132 | |||
FQ221666.1 | - | 2 | 131 | |||
FQ221830.1 | - | 2 | 132 | |||
FQ221978.1 | - | 2 | 132 | |||
FQ221986.1 | - | 2 | 131 | |||
FQ221995.1 | - | 2 | 132 | |||
FQ222044.1 | - | 2 | 128 | |||
FQ222154.1 | - | 2 | 130 | |||
FQ222265.1 | - | 2 | 131 | |||
FQ222417.1 | - | 2 | 128 | |||
FQ222458.1 | - | 2 | 132 | |||
FQ222536.1 | - | 2 | 131 | |||
FQ222629.1 | - | 2 | 128 | |||
FQ222759.1 | - | 2 | 132 | |||
FQ222997.1 | - | 2 | 132 | |||
FQ223042.1 | - | 2 | 131 | |||
FQ223180.1 | - | 2 | 131 | |||
FQ223244.1 | - | 2 | 128 | |||
FQ224329.1 | - | 2 | 128 | |||
FQ224364.1 | - | 2 | 132 | |||
FQ224436.1 | - | 2 | 128 | |||
FQ228463.1 | - | 2 | 131 | |||
FQ228512.1 | - | 2 | 128 | |||
FQ228892.1 | - | 2 | 135 | |||
X06483.1 | - | 1 | 86 |