Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp36l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017172.1 | NP_058868.1 | 2 | 187 | ||
GenBank mRNA | X52590.1 | - | 2 | 187 |