Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144063.1 | NP_001137535.1 | 4, 5 | 466 | ||
NM_020896.3 | NP_065947.1 | 4, 5 | 466 | |||
NM_145638.2 | NP_663613.1 | 4, 5 | 459 | |||
XM_011519873.2 | XP_011518175.1 | 4, 5 | 805 | |||
XM_017017163.1 | XP_016872652.1 | 4, 5 | 813 | |||
GenBank mRNA | AB074006.1 | - | 4, 5 | 407 | ||
AF392453.1 | - | 4, 5 | 379 | |||
AF410430.1 | - | 3, 4 | 324 | |||
AK056510.1 | - | 4, 5 | 424 | |||
AK293000.1 | - | 4, 5 | 415 | |||
AK297475.1 | - | 4, 5 | 414 | |||
AK301003.1 | - | 2, 3 | 225 | |||
AK303205.1 | - | 4, 5 | 284 | |||
AK309411.1 | - | 4, 5 | 444 | |||
AL136918.1 | - | 4, 5 | 379 | |||
BC032646.1 | - | 4, 5 | 380 | 5518491 | ||
BC039579.1 | - | 4, 5 | 377 | 5769002 | ||
DC400863.1 | - | 4, 5 | 466 |