Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019318.1 | NP_062191.1 | 1 | 278 | ||
XM_006255693.2 | XP_006255755.1 | 1 | 285 | |||
XM_006255696.2 | XP_006255758.1 | 1 | 285 | |||
GenBank mRNA | U56242.1 | - | 1 | 278 |