Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021261.2 | NP_067084.1 | 2 | 161 | ||
XM_006236692.2 | XP_006236754.1 | 2 | 212 | |||
GenBank mRNA | BC126092.1 | - | 2 | 130 | ||
CF977046.1 | - | 2 | 165 | |||
FQ221466.1 | - | 2 | 159 | |||
FQ223015.1 | - | 2 | 159 | |||
FQ224120.1 | - | 2 | 161 | |||
FQ224544.1 | - | 2 | 159 | |||
FQ229044.1 | - | 2 | 160 | |||
M17698.1 | - | 2 | 140 | |||
M58404.1 | - | 1 | 235 | |||
M58405.1 | - | 2 | 142 |