Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012635.1 | NP_036767.1 | 3, 4 | 466 | ||
GenBank mRNA | FQ226369.1 | - | 3, 4 | 467 | ||
FQ227841.1 | - | 3, 4 | 467 | |||
FQ233648.1 | - | 3, 4 | 467 | |||
FQ235029.1 | - | 3, 4 | 467 | |||
V01273.1 | - | 3, 4 | 466 |