Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021127.2 | NP_066950.1 | 2 | 315 | ||
GenBank mRNA | AK311943.1 | - | 2 | 280 | ||
BC013120.1 | - | 2 | 238 | 3858958 | ||
BC032663.1 | - | 3 | 421 | 5529754 | ||
BF667112.1 | - | 316 | ||||
D90070.1 | - | 2 | 270 | |||
KU178263.1 | - | 3 | 248 | |||
KU178264.1 | - | 2 | 97 |