Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013104.3 | NP_037236.1 | 1 | 250 | ||
GenBank mRNA | BC099742.1 | - | 1 | 262 | 7937967 | |
FQ220221.1 | - | 1 | 320 | |||
FQ220764.1 | - | 1 | 323 | |||
FQ230155.1 | - | 1 | 326 |