Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270688.1 | NP_001257617.1 | 3 | 122 | ||
NM_001270689.1 | NP_001257618.1 | 3 | 150 | |||
NM_021909.2 | NP_068709.2 | 3 | 226 | |||
GenBank mRNA | BC060571.1 | - | 3 | 114 | 6891214 | |
FM112735.1 | - | 3 | 241 | |||
FM122563.1 | - | 3 | 100 | |||
FQ220058.1 | - | 3 | 150 |