Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022196.2 | NP_071532.2 | 2 | 181 | ||
XM_006251364.2 | XP_006251426.1 | 3 | 316 | |||
XM_008770359.1 | XP_008768581.1 | 2 | 886 | |||
GenBank mRNA | AB010275.1 | - | 2 | 181 | ||
M32748.1 | - | 2 | 181 |