Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023990.1 | NP_076480.1 | 1 | 615 | ||
GenBank mRNA | AB030833.1 | - | 1 | 615 |