Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024162.1 | NP_077076.1 | 1, 2 | 110 | ||
GenBank mRNA | FQ217465.1 | - | 1, 2 | 114 | ||
J02773.1 | - | 1, 2 | 110 | |||
M18034.1 | - | 1, 2 | 97 |