Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021240.3 | NP_067063.1 | 2 | 546 | ||
GenBank mRNA | AF193873.1 | - | 2 | 127 | ||
AF217741.2 | - | 1 | 262 | |||
BC113584.1 | - | 2 | 546 | 8322636 | ||
BC117245.1 | - | 2 | 546 | 40125796 |