Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031116.3 | NP_112378.2 | 2 | 200 | ||
GenBank mRNA | BC058138.1 | - | 2 | 216 | 6921268 | |
EF121972.1 | - | 158 | ||||
EF122007.1 | - | 2 | 158 | |||
U06436.1 | - | 2 | 158 |