Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031590.1 | NP_113778.1 | 4 | 600 | ||
GenBank mRNA | AF259981.1 | - | 4 | 600 | ||
BC089187.1 | - | 3 | 410 |