Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marcksl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030862.2 | NP_110489.1 | 2 | 475 | ||
GenBank mRNA | AJ301677.1 | - | 2 | 368 | ||
BC081789.1 | - | 2 | 475 | 7134056 |
Gene Symbol: LOC102550530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759283.1 | XP_008757505.1 | 2 | 434 |
Gene Symbol: LOC103689966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759211.1 | XP_008757433.1 | 1 | 509 |