Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053800.3 | NP_446252.1 | 3 | 210 | ||
GenBank mRNA | AF311055.1 | - | 3 | 210 | ||
BC058454.1 | - | 295 | 6886988 | |||
FQ209943.1 | - | 292 | ||||
FQ210839.1 | - | 295 | ||||
FQ216849.1 | - | 321 | ||||
FQ217822.1 | - | 290 | ||||
FQ221646.1 | - | 263 | ||||
X14878.1 | - | 270 |