Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nip7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138847.1 | NP_620202.1 | 2 | 161 | ||
GenBank mRNA | AF158186.1 | - | 2 | 161 | ||
BC059114.1 | - | 2 | 132 | 6921568 | ||
BC086589.1 | - | 2 | 84 | 7309991 |