Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138841.1 | NP_620196.1 | 2 | 757 | ||
GenBank mRNA | AF208288.1 | - | 2 | 757 |
Set Membership: |
G-protein coupled receptor |