Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175599.1 | NP_783189.1 | 1 | 94 |
Gene Symbol: LOC100910174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748672.3 | XP_003748720.1 | 1 | 86 |
Gene Symbol: Taar8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227718.2 | XP_006227780.2 | 2 | 70 | ||
XM_006227719.2 | XP_006227781.1 | 2 | 492 | |||
XM_008758615.1 | XP_008756837.1 | 3 | 650 |
Gene Symbol: Taar8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758669.1 | XP_008756891.1 | 1 | 94 |
Set Membership: |
G-protein coupled receptor |