Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017312.2 | NP_059008.1 | 3 | 338 | ||
GenBank mRNA | AF027954.1 | - | 2 | 248 | ||
BC078871.1 | - | 3 | 338 | 7132448 | ||
FQ227820.1 | - | 2 | 179 | |||
HQ696784.1 | - | 3 | 456 |