Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019323.1 | NP_062196.1 | 252 | |||
GenBank mRNA | U67912.1 | - | 233 | |||
U72143.1 | - | 252 |
Gene Symbol: Mcpt8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135010.1 | NP_001128482.1 | 3 | 269 | ||
GenBank mRNA | FQ229939.1 | - | 3 | 331 |