Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301064.1 | NP_001287993.1 | 10 | 998 | ||
NM_018427.4 | NP_060897.3 | 11 | 1088 | |||
XM_005255376.1 | XP_005255433.1 | 9 | 863 | |||
XM_005255377.4 | XP_005255434.1 | 8 | 812 | |||
XM_011522540.2 | XP_011520842.1 | 6 | 500 | |||
XM_011522541.1 | XP_011520843.1 | 10 | 927 | |||
GenBank mRNA | AF227156.1 | - | 11 | 1054 | ||
AJ272050.1 | - | 11 | 1008 | |||
AK225662.1 | - | 11 | 1064 | |||
AK295486.1 | - | 8 | 681 | |||
AK300253.1 | - | 10 | 907 | |||
AK302994.1 | - | 8 | 735 | |||
AK303405.1 | - | 7 | 581 | |||
AK304856.1 | - | 11 | 954 | |||
AK314769.1 | - | 11 | 1002 | |||
AK316520.1 | - | 10 | 912 | |||
BC036182.1 | - | 11 | 1003 | |||
BC071868.1 | - | 3 | 323 | |||
BC132688.1 | - | 11 | 1005 | 40146710 | ||
BC132690.1 | - | 11 | 1005 | 40146712 | ||
DA764092.1 | - | 3 | 291 |