Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022214.1 | NP_071550.1 | 1, 2 | 111 | ||
XM_008770053.1 | XP_008768275.1 | 8, 9 | 1508 | |||
XM_008770054.1 | XP_008768276.1 | 1, 2 | 528 | |||
XM_008770055.1 | XP_008768277.1 | 8, 9 | 1505 | |||
GenBank mRNA | U90448.1 | - | 1, 2 | 111 |