Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022697.2 | NP_073188.2 | 1, 2 | 103 | ||
XM_006228288.2 | XP_006228350.1 | 2, 3 | 201 | |||
GenBank mRNA | BC081800.1 | - | 1, 2 | 103 | 7131175 | |
FQ209745.1 | - | 2, 3 | 128 | |||
FQ212339.1 | - | 2, 3 | 129 | |||
FQ217837.1 | - | 2, 3 | 130 | |||
FQ221737.1 | - | 2, 3 | 125 | |||
FQ221950.1 | - | 1, 2 | 94 | |||
FQ222287.1 | - | 2, 3 | 125 | |||
FQ222709.1 | - | 2, 3 | 121 | |||
FQ223000.1 | - | 2, 3 | 129 | |||
FQ223241.1 | - | 2, 3 | 124 | |||
FQ223819.1 | - | 2, 3 | 125 | |||
FQ224137.1 | - | 2, 3 | 129 | |||
FQ226808.1 | - | 1, 2 | 447 | |||
FQ227480.1 | - | 2, 3 | 129 | |||
FQ228328.1 | - | 2, 3 | 133 | |||
FQ228578.1 | - | 2, 3 | 128 | |||
X52619.1 | - | 2, 3 | 124 |