Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022952.2 | NP_075241.2 | 2 | 141 | ||
GenBank mRNA | BC088138.1 | - | 2 | 141 | 7374432 | |
FQ234449.1 | - | 2 | 160 | |||
M37194.1 | - | 2 | 143 | |||
U75917.1 | - | 2 | 160 |