Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024131.1 | NP_077045.1 | 2 | 317 | ||
GenBank mRNA | FQ209890.1 | - | 2 | 334 | ||
FQ209898.1 | - | 2 | 289 | |||
FQ210467.1 | - | 2 | 290 | |||
FQ210521.1 | - | 2 | 333 | |||
FQ220991.1 | - | 2 | 327 | |||
Z36980.1 | - | 2 | 317 |