Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024135.2 | NP_077049.2 | 3 | 241 | ||
XM_006251236.2 | XP_006251298.1 | 2 | 1558 | |||
XM_008770275.1 | XP_008768497.1 | 3 | 344 | |||
XM_008770276.1 | XP_008768498.1 | 2 | 1375 | |||
XM_008770277.1 | XP_008768499.1 | 2 | 1558 | |||
XM_008770278.1 | XP_008768500.1 | 3 | 343 | |||
XM_008770279.1 | XP_008768501.1 | 3 | 343 | |||
GenBank mRNA | BC065578.1 | - | 2 | 426 | 5623751 | |
D31874.1 | - | 3 | 241 | |||
D31875.1 | - | 2 | 470 | |||
D31876.1 | - | 2 | 470 | |||
D31877.1 | - | 2 | 470 |