Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031110.1 | NP_112372.1 | 2 | 89 | ||
GenBank mRNA | BC058465.1 | - | 2 | 157 | 6920362 | |
FQ210967.1 | - | 2 | 124 | |||
FQ217176.1 | - | 2 | 112 | |||
FQ221188.1 | - | 2 | 165 | |||
FQ221414.1 | - | 2 | 116 | |||
FQ221793.1 | - | 2 | 112 | |||
FQ221872.1 | - | 2 | 112 | |||
FQ221910.1 | - | 2 | 119 | |||
FQ222283.1 | - | 2 | 141 | |||
FQ222285.1 | - | 2 | 119 | |||
FQ222811.1 | - | 2 | 113 | |||
FQ222934.1 | - | 2 | 144 | |||
FQ222947.1 | - | 2 | 112 | |||
FQ223175.1 | - | 2 | 112 | |||
FQ223203.1 | - | 2 | 114 | |||
FQ223895.1 | - | 2 | 112 | |||
FQ229022.1 | - | 2 | 112 | |||
K03250.1 | - | 2 | 89 |
Gene Symbol: RGD1563352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_575471.4 | XP_575471.1 | 1 | 87 |