Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053507.1 | NP_445959.1 | 2 | 129 | ||
XM_008767598.1 | XP_008765820.1 | 2 | 241 | |||
GenBank mRNA | AY017337.1 | - | 2 | 129 |