Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053669.1 | NP_446121.1 | 2 | 463 | ||
XM_006249092.2 | XP_006249154.1 | 2 | 1215 | |||
XM_006249093.2 | XP_006249155.1 | 2 | 1216 | |||
XM_008769112.1 | XP_008767334.1 | 3 | 1014 | |||
XM_008769113.1 | XP_008767335.1 | 2 | 478 | |||
GenBank mRNA | AF095576.1 | - | 2 | 463 |