Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147211.1 | NP_671744.1 | 3 | 309 | ||
XM_008762874.1 | XP_008761096.1 | 3 | 744 | |||
XM_008762875.1 | XP_008761097.1 | 3 | 3071 | |||
XM_008762876.1 | XP_008761098.1 | 3 | 744 | |||
XM_008762877.1 | XP_008761099.1 | 3 | 742 | |||
GenBank mRNA | U25281.1 | - | 3 | 309 | ||
U31159.1 | - | 3 | 309 |