Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172065.2 | NP_742062.1 | 1, 3 | 92 | ||
GenBank mRNA | L17318.1 | - | 1, 3 | 93 |