Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007379.2 | NP_031405.2 | 2, 3 | 260 | ||
GenBank mRNA | AK169438.1 | - | 2, 3 | 260 | ||
X75927.2 | - | 2, 3 | 164 |