Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077190.2 | NP_001070658.1 | 3 | 588 | ||
NM_001077192.2 | NP_001070660.1 | 3 | 588 | |||
NM_001077193.2 | NP_001070661.1 | 3 | 588 | |||
NM_007380.3 | NP_031406.2 | 3 | 588 | |||
NM_145994.2 | NP_666106.1 | 3 | 588 | |||
GenBank mRNA | AF420251.1 | - | 3 | 524 | ||
AK034476.1 | - | 3 | 470 | |||
AK151026.1 | - | 3 | 496 | |||
AK151363.1 | - | 3 | 567 | |||
AK152046.1 | - | 1 | 508 | |||
AK152061.1 | - | 3 | 452 | |||
AK152184.1 | - | 3 | 452 | |||
AK153130.1 | - | 3 | 496 | |||
AK164040.1 | - | 3 | 504 | |||
AK165924.1 | - | 3 | 561 | |||
AK167328.1 | - | 3 | 547 | |||
AK167568.1 | - | 3 | 543 | |||
AK167744.1 | - | 3 | 535 | |||
AK181529.1 | - | 1 | 116 | |||
AK184875.1 | - | 1 | 32 | |||
AK190191.1 | - | 1 | 32 | |||
AK199630.1 | - | 1 | 32 | |||
AK201770.1 | - | 1 | 32 | |||
AK215001.1 | - | 1 | 32 | |||
AY033645.1 | - | 3 | 432 | |||
BC004657.1 | - | 3 | 539 | 3498068 | ||
BC079642.1 | - | 3 | 494 | 6816252 | ||
U17698.1 | - | 2 | 214 |