Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007399.3 | NP_031425.2 | 2 | 312 | ||
GenBank mRNA | AF011379.1 | - | 2 | 133 | ||
BC057346.1 | - | 2 | 274 | |||
BC066207.1 | - | 2 | 317 |