Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089086.1 | - | 4, 5 | 326 | ||
AK089169.1 | - | 4, 5 | 348 | |||
AK150411.1 | - | 4, 5 | 328 | |||
AK152675.1 | - | 4, 5 | 345 | |||
AK152769.1 | - | 4, 5 | 327 | |||
AK153065.1 | - | 4, 5 | 327 | |||
AK155772.1 | - | 4, 5 | 326 | |||
AK155912.1 | - | 4, 5 | 346 | |||
AK160041.1 | - | 4, 5 | 329 | |||
AK170019.1 | - | 4, 5 | 351 | |||
AK171022.1 | - | 4, 5 | 348 | |||
AK171060.1 | - | 4, 5 | 326 | |||
BC025584.1 | - | 4, 5 | 380 | 5324943 | ||
X13335.1 | - | 4, 5 | 325 |