Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243013.1 | NP_001229942.1 | 6 | 1509 | ||
NM_001243014.1 | NP_001229943.1 | 6 | 1509 | |||
NM_019624.3 | NP_062570.1 | 6 | 1509 | |||
NM_019625.3 | NP_062571.1 | 6 | 1509 | |||
NM_203444.3 | NP_982269.2 | 6 | 1509 | |||
XM_005253558.2 | XP_005253615.1 | 6 | 1449 | |||
XM_011538095.2 | XP_011536397.1 | 6 | 1732 | |||
XM_011538096.2 | XP_011536398.1 | 6 | 1528 | |||
XM_011538098.2 | XP_011536400.1 | 5 | 2374 | |||
XM_011538099.2 | XP_011536401.1 | 5 | 1057 | |||
XM_017019103.1 | XP_016874592.1 | 7 | 1570 | |||
XM_017019104.1 | XP_016874593.1 | 5 | 1057 | |||
GenBank mRNA | AB040953.2 | - | 6 | 4038 | ||
AB045381.2 | - | 5 | 1258 | |||
AB112582.1 | - | 6 | 1460 | |||
AB112583.1 | - | 6 | 1460 | |||
AB177852.1 | - | 6 | 1421 | |||
AF216494.1 | - | 6 | 1497 | |||
AK027624.1 | - | 6 | 1388 | |||
AK074738.1 | - | 3 | 453 | |||
AK304295.1 | - | 6 | 1505 | |||
AL833963.1 | - | 5 | 625 | |||
BC017348.2 | - | 5 | 1224 | 5014247 | ||
BC064384.1 | - | 6 | 1777 | 5744451 |