Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007435.1 | NP_031461.1 | 1 | 1049 | ||
GenBank mRNA | AK088792.1 | - | 1 | 919 | ||
AK089318.1 | - | 1 | 477 | |||
AK153589.1 | - | 1 | 897 | |||
AK170424.1 | - | 1 | 477 | |||
BC011273.1 | - | 1 | 868 | 3710503 | ||
BC079840.1 | - | 1 | 927 | 6810147 | ||
Z33637.1 | - | 1 | 1049 |