Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051219.1 | - | 1 | 266 | ||
AK078243.1 | - | 266 | ||||
AK079571.1 | - | 1 | 247 | |||
AK079639.1 | - | 1 | 247 | |||
AK086262.1 | - | 1 | 248 | |||
AK140932.1 | - | 1 | 286 | |||
AK148462.1 | - | 1 | 248 | |||
AK152156.1 | - | 1 | 284 | |||
AK152735.1 | - | 1 | 284 | |||
AK152800.1 | - | 1 | 284 | |||
AK159246.1 | - | 1 | 306 | |||
AK163040.1 | - | 1 | 247 | |||
AK169157.1 | - | 1 | 247 | |||
AK170195.1 | - | 1 | 267 | |||
AK170251.1 | - | 1 | 247 | |||
BC003797.1 | - | 1 | 208 | 3490519 | ||
U14390.1 | - | 1 | 238 |