Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007445.2 | NP_031471.2 | 2 | 482 | ||
GenBank mRNA | AY911505.1 | - | 2 | 482 | ||
BC150154.1 | - | 2 | 481 | |||
BC150477.1 | - | 2 | 481 |